Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LGALS1 Mouse anti-Human, Clone: 24CT661.4.3, Abnova™

Mouse monoclonal antibody raised against human LGALS1.

Manufacturer:  Abnova Corporation MAB12663

Catalog No. 89-934-474


Add to cart

Description

Description

The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. This gene product may act as an autocrine negative growth factor that regulates cell proliferation. [provided by RefSeq]

Specifications

Specifications

LGALS1
24CT661.4.3
Mouse monoclonal antibody raised against human LGALS1.
In ascites (0.09% sodium azide).
DKFZp686E23103/GAL1/GBP
Mouse
IgG1
RUO
Primary
3956
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Liquid
LGALS1
Purified His-tagged GLT protein.
50 uL
Store at 4°C for six months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit