Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LGALS13 Mouse anti-Human, Clone: PP13/1165, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14808

Catalog No. 89-034-891


Add to cart

Description

Description

Mouse monoclonal antibody raised against partial recombinant human LGALS13.

  • High specificity
  • High reactivity
Specifications

Specifications

LGALS13
PP13/1165
Mouse monoclonal antibody raised against partial recombinant human LGALS13.
In 1 mM PBS.
Liquid
LGALS13
Recombinant protein corresponding to amino acids 23-134 of human LGALS13.
Protein A/G purification
RUO
Primary
29124
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
ELISA Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Q9UHV8
GAL13|PLAC8|PP13
Mouse
IgG2b
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)