Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LGALS13 Polyclonal Antibody
Rabbit Polyclonal Antibody
$178.40 - $511.50
Specifications
Antigen | LGALS13 |
---|---|
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119347
|
Invitrogen™
PA5119347 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121797
|
Invitrogen™
PA5121797 |
20 μL |
Each for $178.40
|
|
|||||
Description
LGALS13 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Lysophospholipases are enzymes that act on biological membranes to regulate the multifunctional lysophospholipids. The protein encoded by this gene has lysophospholipase activity. It is composed of two identical subunits which are held together by disulfide bonds. This protein has structural similarity to several members of the beta-galactoside-binding S-type lectin family.Specifications
LGALS13 | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
LGALS13 | |
beta-galactoside-binding lectin; GAL13; gal-13; Galactoside-binding soluble lectin 13; galectin 13; galectin-13; lectin, galactoside binding soluble 13; lectin, galactoside-binding, soluble, 13; LGALS13; PLAC8; placental protein 13; placental tissue protein 13; PP13 | |
LGALS13 | |
Primary | |
Antigen affinity chromatography |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9UHV8 | |
29124 | |
Recombinant protein Placental Protein 13. The antigen corresponds to amino acid range 1-139 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title