Learn More
LGALS13 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 178231AP
Catalog No. 50-173-1818
Add to cart
Description
Description
Lysophospholipases are enzymes that act on biological membranes to regulate the multifunctional lysophospholipids. The protein encoded by this gene has lysophospholipase activity. It is composed of two identical subunits which are held together by disulfide bonds. This protein has structural similarity to several members of the beta-galactoside-binding S-type lectin family.
Specifications
Specifications
LGALS13
0.44 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UHV8
Gal 13, GAL13, Galectin 13, LGALS13, PLAC8, Placental protein 13, Placental tissue protein 13, PP13
Rabbit
IgG
150 μL
-20°C
Primary
29124
Western Blot
Unconjugated
LGALS13
Liquid
LGALS13
LGALS13 Fusion Protein Ag12192
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human