Learn More
LGALS9 Mouse anti-Human, Clone: OTI8B11, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF805615
Catalog No. 50-166-7505
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The protein encoded by this gene is an S-type lectin. It is overexpressed in Hodgkin ft.s disease tissue and might participate in the interaction between the H&RS cells with their surrounding cells and might thus play a role in the pathogenesis of this disease and/or its associated immunodeficiency. Multiple alternatively spliced transcript variants have been found for this gene.
Specifications
Specifications
LGALS9
Monoclonal
Unconjugated
LGALS9
Lyophilized
LGALS9
Full length human recombit protein of human LGALS9 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI8B11
PBS with 8% trehalose and no preservative; pH 7.3
O00182
HUAT, LGALS9A
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
3965