Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lgi1, Mouse anti-Mouse, ATTO633, Clone: S283-7, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB18616

Catalog No. 89-338-541


Add to cart

Description

Description

Specifications

Specifications

Lgi1
S283-7
Immunocytochemistry Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide).
BB130740
Lgi1
Recombinant protein corresponding to amino acids 37-113 at LRRNT domain and first LRR repeat of mouse Lgi1.
Monoclonal
100 μg
Store at -20°C.
Antibody
Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Western Blot
ATTO633
Liquid
leucine-rich repeat LGI family, member 1
56839
Mouse
IgG2a
Primary
RUO
Human,Mouse,Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit