Learn More
Lgi1 Mouse anti-Human, Mouse, Rat, DyLight 488, Clone: S283-7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242190G
Catalog No. NBP242190G
Add to cart
Description
Description
Lgi1 Monoclonal antibody specifically detects Lgi1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
Lgi1
S283-7
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
LGI1
Fusion protein amino acids 37-113 (LRRNT domain and first LRR repeat) of mouse LGI1. Rat: 100% identity (77/77 amino acids identical). Human: 98% identity (76/77 amino acids identical). ~50% identity with LGI2, LGI3 and LGI4.
Protein G purified
Neuroscience
Primary
9211
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
epilepsy, partial, EPITEMPIN, Epitempin-1, EPTleucine-rich glioma-inactivated protein 1, ETL1epitempin-1, IB1099, leucine-rich, glioma inactivated 1
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
approx 50% identity with LGI2, LGI3 and LGI4.Detects approx 60kDa.