Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lgi1 Antibody (S283-7), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242190H
Description
Lgi1 Monoclonal specifically detects Lgi1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Lgi1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
LGI1 | |
Fusion protein amino acids 37-113 (LRRNT domain and first LRR repeat) of mouse LGI1. Rat: 100% identity (77/77 amino acids identical). Human: 98% identity (76/77 amino acids identical). ∼50% identity with LGI2, LGI3 and LGI4. | |
0.1 mL | |
Primary | |
approx 50% identity with LGI2, LGI3 and LGI4.Detects approx 60kDa. | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Immunohistochemistry | |
S283-7 | |
Western Blot, Immunohistochemistry | |
epilepsy, partial, EPITEMPIN, Epitempin-1, EPTleucine-rich glioma-inactivated protein 1, ETL1epitempin-1, IB1099, leucine-rich, glioma inactivated 1 | |
Mouse | |
Protein G purified | |
Neuroscience | |
9211 | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction