Learn More
LGP Consulting HCTS2000 MK2 Tube Sorter
Quickly and efficiently sorts up to 2000 sample tubes per hour
Manufacturer:  LGP ConsultingSupplier Diversity Partner 30 104933
 View more versions of this product
Catalog No. 23-047-105
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Sorting closed primary sample tubes used in medical laboratories
5 additional target bins
160kg
56 in.
HCTS2000 MK2 tube sorter
MK2 Tube Sorter with Warning Light
351 lb.
143cm