Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LGP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | LGP2 |
---|---|
Concentration | 0.17 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-7087
![]() ![]() |
Proteintech Group Inc
113551AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Anti-viral innate immune responses are triggered by pathogen-associated molecular patters (such as the accumulation of intracellular nucleic acids resulting from virus infections) and represent the first line of defense against numerous infectious organisms. The Toll-like receptors (TLR) 3, 7, 8, and 9 are expressed in immune cells and function as transmembrane pattern recognition receptors to detect foreign nucleic acids. Other proteins that play similar roles, such as RIG-1 and MDA5 are members of a CARD-helicase family and are expressed in the cytoplasm. A third protein, LGP2, is similar to RIG-1 and MDA5, except for lacking a homologous CARD domain. It is thought to act as an element of negative-feedback regulation of intracellular antiviral signaling. At least four isoforms of LGP2 are known to exist.Specifications
LGP2 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q96C10, Q99J87 | |
303538, 79132, 80861 | |
LGP2 Fusion Protein Ag1910 | |
Primary | |
-20°C | |
DHX58 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
D11LGP2, D11lgp2e, DHX58, LGP2, Protein D11Lgp2 homolog, RIG I like receptor LGP2, RLR | |
Dhx58 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title