Learn More
Lgr4/GPR48 Mouse anti-Human, Alexa Fluor 750, Clone: 852229, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB7750S100UG
Catalog No. FAB7750S100
Add to cart
This item is not returnable. View return policy
Description
Description
Lgr4/GPR48 Monoclonal antibody specifically detects Lgr4/GPR48 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Lgr4/GPR48
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q9BXB1
LGR4
NS0 mouse myeloma cell line transfected with human Lgr4/GPR48 Met1-Asp951 Accession # Q9BXB1
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human Lgr4/GPR48 in Lgr4-transfected cell-based ELISA. Stains human Lgr4/GPR48 transfected but not irrelevant transfected cells in flow cytometry.
Flow Cytometry
852229
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
GPR48, GPR48G protein-coupled receptor 48, G-protein coupled receptor 48, leucine-rich repeat containing G protein-coupled receptor 4
Mouse
IgG2b
RUO
Primary
55366
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)