Learn More
Lgr4/GPR48 Mouse anti-Human, APC, Clone: 852229, R&D Systems
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB7750A100
Catalog No. FAB7750A100
Add to cart
This item is not returnable. View return policy
Description
Description
Lgr4/GPR48 Monoclonal antibody specifically detects Lgr4/GPR48 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Lgr4/GPR48
852229
Flow Cytometry 10 μL/106 cells
Q9BXB1
GPR48, GPR48G protein-coupled receptor 48, G-protein coupled receptor 48, leucine-rich repeat containing G protein-coupled receptor 4
Mouse
IgG2b
100 Tests
Primary
55366.0
Human
Flow Cytometry
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified, Supernatant, Conjugated
LGR4
NS0 mouse myeloma cell line transfected with human Lgr4/GPR48, Met1-Asp951, Accession No. Q9BXB1
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human Lgr4/GPR48 in Lgr4-transfected cell-based ELISA. Stains human Lgr4/GPR48 transfected but not irrelevant transfected cells in flow cytometry.