Learn More
Lgr5/GPR49 Mouse anti-Human, Alexa Fluor 647, Clone: 707042, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8078R100UG
Catalog No. FAB8078R100
Add to cart
This item is not returnable. View return policy
Description
Description
Published as Clone RD42
Specifications
Specifications
Lgr5/GPR49
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
O75473
LGR5
Chinese hamster ovary cell line CHO-derived recombinant human Lgr5/GPR49 Met1-Ile560 Accession # O75473
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human Lgr5/GPR49 in ELISA. Stains human and mouse Lgr5 transfected cells but not irrelevant transfectants in Flow Cytometry and Immunocytochemistry. This antibody, also known as “RD42”, has been found to detect an epitope in C-terminal LRR cap of LGR5 (Ref. 1).
Flow Cytometry
707042
Alexa Fluor 647
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
FEX, G protein-coupled receptor 49, GPR49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38
Mouse
IgG2a
RUO
Primary
8549
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)