Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lgr5/GPR49 Rat anti-Mouse, Alexa Fluor 647, Clone-803420, R&D Systems™

Rat Monoclonal Antibody

Manufacturer:  R&D Systems FAB8240R100UG

 View more versions of this product

Catalog No. FAB8240R100


Add to cart

Description

Description

Flow Cytometry: Please use 0.25-1 ug of conjugated antibody per 10e6 cells.
Specifications

Specifications

Lgr5/GPR49
803420
Alexa Fluor 647
Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
FEX, G protein-coupled receptor 49, GPR49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38
Rat
IgG2b
100 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects mouse Lgr5/GPR49 in direct ELISAs. Stains mouse Lgr5 transfected cells but not irrelevant transfectants in flow cytometry. This clone, also known as “RD20”, has been found to detect an epitope in C-terminal LRR cap of LGR5 (Ref. 1).
Flow Cytometry
0.2 mg/mL
Flow Cytometry 0.1 ug/10^6 cells
Q9Z1P4
LGR5
E. coli-derived recombinant mouse Lgr5/GPR49, Lys375-Pro547, Accession # Q9Z1P4
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
8549
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit