Learn More
LGR5 Mouse anti-Human, Clone: OTI11C2, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF808304
Catalog No. 50-166-8762
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
G protein-coupled receptor GPR49 has been reported to be expressed in brain, skeletal muscle, placenta, and spinal cord. ESTs have been isolated from normal human brain (amygdala), embryo, and placenta libraries and from human germ cell, uterus, and brain libraries. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This ""turn-off"" activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Two blockbuster drugs which have been developed as GPCR-targeted pharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.
Specifications
Specifications
LGR5
Monoclonal
Unconjugated
LGR5
Lyophilized
LGR5
Human recombit protein fragment corresponding to amino acids 22-543 of human LGR5 produced in HEK293 cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI11C2
PBS with 8% trehalose and no preservative; pH 7.3
O75473
FEX, GPR49, GPR67, GRP49, HG38
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
8549