Learn More
Lgr6 Mouse anti-Human, Alexa Fluor 488, Clone: 918719, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8458G100UG
Catalog No. FAB8458G100
Add to cart
This item is not returnable. View return policy
Description
Description
Lgr6 Monoclonal antibody specifically detects Lgr6 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Lgr6
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q9HBX8
LGR6
NS0 mouse myeloma cell line transfected with human Lgr6 Met1-Val967 Accession # Q9HBX8
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Stains human Lgr6 transfectants but not irrelevant transfectants in flow cytometry.
Flow Cytometry
918719
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
FLJ14471, gonadotropin receptor, GPCR, leucine-rich repeat containing G protein-coupled receptor 6, leucine-rich repeat-containing G protein-coupled receptor 6, leucine-rich repeat-containing G-protein coupled receptor 6, VTS20631
Mouse
IgG1
RUO
Primary
59352
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)