Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LGR6 Mouse anti-Human, Clone: OTI2A3, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504757

Catalog No. 50-167-5374


Add to cart

Description

Description

LGR6 has been reported to be expressed in rat adrenal, brain, colon, heart, kidney, ovary, oviduct, small intestine, spleen, testis, and uterus. ESTs have been isolated from human B-cell/lung/testis, blood, brain, colon, eye, germ cell, head/neck, heart, heart/melanocyte/uterus, lung, ovary, stomach, testis, and uterus libraries.
Specifications

Specifications

LGR6
Monoclonal
2.03 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9HBX8
GPCR, VTS20631
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
59352
Immunocytochemistry, Immunofluorescence, Western Blot
OTI2A3
Unconjugated
LGR6
Liquid
LGR6
Human recombit protein fragment corresponding to amino acids 25-250 of human LGR6 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit