Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Thermo Scientific™ LH-4000W 8 x 500mL Windshielded Swinging Bucket Rotor
Support a diverse array of sample processing applications without the need to modify existing protocols.
Manufacturer:  Thermo Scientific™ 75006436
 View more versions of this product
Catalog No. 75-006-436
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Support a diverse array of sample processing applications without the need to modify existing protocols with the Thermo Scientific™ LH-4000W 8 x 500mL Windshield Swinging Bucket Rotor. Choose from round, DoubleSpin™ and Double blood bag buckets with a wide array of available adapters, providing capacity flexibility ranging from 1.5 to 250mL.
Ordering Information:
Buckets, biocontainment lids and DoubleSpin buckets are sold separately; order rotor body (75006476) and components to complete assembly. Tube adapters and microplate adapter for DoubleSpin bucket are also available separately. Contact your Sales Representative for information.
Compatible with:
Thermo Scientific™ Heraeus™ Cryofuge 5500i series centrifuges
Description & Specifications
Specifications
Thermo Scientific™ Sorvall™ RC 4 Series, Heraeus™ Cryofuge 5500i and Heraeus™ Multifuge™ 4 large capacity centrifuges
----
Double Blood Bag Bucket