Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LH 846, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
639052-78-1
C16H13ClN2OS
316.803
DYHAMRNAHTWYKY-UHFFFAOYSA-N
n-5-chloro-6-methyl-1,3-benzothiazol-2-yl-2-phenylacetamide,benzeneacetamide, n-5-chloro-6-methyl-2-benzothiazolyl,n-5-chloro-6-methyl-2-benzothiazolyl benzeneacetamide,n-5-chloro-6-methylbenzothiazol-2-yl-2-phenylacetamide,n-5-chloro-6-methyl-benzothiazol-2-yl-2-phenyl-acetamide,n-5-chloro-6-methylbenzo d thiazol-2-yl-2-phenylacetamide
851474
N-(5-chloro-6-methyl-1,3-benzothiazol-2-yl)-2-phenylacetamide
CC1=CC2=C(C=C1Cl)N=C(S2)NC(=O)CC3=CC=CC=C3
This item is not returnable. View return policy
639052-78-1
C16H13ClN2OS
316.803
DYHAMRNAHTWYKY-UHFFFAOYSA-N
n-5-chloro-6-methyl-1,3-benzothiazol-2-yl-2-phenylacetamide,benzeneacetamide, n-5-chloro-6-methyl-2-benzothiazolyl,n-5-chloro-6-methyl-2-benzothiazolyl benzeneacetamide,n-5-chloro-6-methylbenzothiazol-2-yl-2-phenylacetamide,n-5-chloro-6-methyl-benzothiazol-2-yl-2-phenyl-acetamide,n-5-chloro-6-methylbenzo d thiazol-2-yl-2-phenylacetamide
851474
N-(5-chloro-6-methyl-1,3-benzothiazol-2-yl)-2-phenylacetamide
CC1=CC2=C(C=C1Cl)N=C(S2)NC(=O)CC3=CC=CC=C3

Selective casein kinase 1δ inhibitor

Selective inhibitor of casein kinase (CK) 1δ (IC50 values are 290 nM, 1.3 μM and 2.5 μM for CK1δ, ε and α); displays no inhibitory activity at CK2. Inhibits CK1δ-dependent phosphorylation and degradation of PER1 protein. Shown to lengthen the circadian period in U2OS cells, with minimal effect on amplitude.
Quantity 50mg
Formula Weight 316.81
Percent Purity >98%
Chemical Name or Material LH 846

Recommended Storage : Store at +4°C