Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LH 846, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective inhibitor of casein kinase (CK) 1δ (IC50 values are 290 nM, 1.3 μM and 2.5 μM for CK1δ, ε and α); displays no inhibitory activity at CK2. Inhibits CK1δ-dependent phosphorylation and degradation of PER1 protein. Shown to lengthen the circadian period in U2OS cells, with minimal effect on amplitude.
Specifications
Quantity | 10 mg |
Formula Weight | 316.81 |
Percent Purity | >98% |
Chemical Name or Material | LH 846 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction