Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LH beta Antibody (L1), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10073010V
Description
LH beta Monoclonal specifically detects LH beta in Human samples. It is validated for Western Blot, Sandwich ELISA.Specifications
| LH beta | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LHB | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified natural LH beta. | |
| 0.2 mL | |
| Primary | |
| High affinity for the beta chain of LH. No cross-reaction with FSH, TSH or HCG. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA | |
| L1 | |
| Western Blot, Sandwich ELISA | |
| CGB4, hLHB, interstitial cell stimulating hormone, beta chain, LSH-B, LSH-beta, luteinizing hormone beta polypeptide, luteinizing hormone beta subunit, lutropin beta chain, lutropin subunit beta | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 3972 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction