Learn More
Abnova Human LHFPL5 Full-length ORF (NP_872354.1, 1 a.a. - 219 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Manufacturer: Abnova H00222662P01L
Description
This gene is a member of the lipoma HMGIC fusion partner (LHFP) gene family, which is a subset of the superfamily of tetraspan transmembrane protein encoding genes. Mutations in this gene result in deafness in humans, and a mutation in a similar gene in mice results in deafness and vestibular dysfunction with severe degeneration of the organ of Corti. It is proposed to function in hair bundle morphogenesis. [provided by RefSeq]
Sequence: MVKLLPAQEAAKIYHTNYVRNSRAVGVMWGTLTICFSVLVMALFIQPYWIGDSVNTPQAGYFGLFSYCVGNVLSSELICKGGPLDFSSIPSRAFKTAMFFVALGMFLIIGSIICFSLFFICNTATVYKICAWMQLAAATGLMIGCLVYPDGWDSSEVRRMCGEQTGKYTLGHCTIRWAFMLAILSIGDALILSFLAFVLGYRQDKLLPDDYKADGTEEVSpecifications
NP_872354.1 | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
50.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DFNB67|MGC33835|TMHS|dJ510O8.8 | |
LHFPL5 | |
Yes |
wheat germ expression system | |
Liquid | |
222662 | |
LHFPL5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVKLLPAQEAAKIYHTNYVRNSRAVGVMWGTLTICFSVLVMALFIQPYWIGDSVNTPQAGYFGLFSYCVGNVLSSELICKGGPLDFSSIPSRAFKTAMFFVALGMFLIIGSIICFSLFFICNTATVYKICAWMQLAAATGLMIGCLVYPDGWDSSEVRRMCGEQTGKYTLGHCTIRWAFMLAILSIGDALILSFLAFVLGYRQDKLLPDDYKADGTEEV | |
RUO | |
LHFPL5 | |
Wheat Germ (in vitro) | |
GST |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.