Learn More
LHX1 Mouse anti-Human, Clone: OTI3A9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504587

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of a large protein family which contains the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator and be involved in control of differentiation and development of neural and lymphoid cells. A similar protein in mice is an essential regulator of the vertebrate head organizer.Specifications
LHX1 | |
Monoclonal | |
Unconjugated | |
LHX1 | |
LIM-1, LIM1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3975 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI3A9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P48742 | |
LHX1 | |
Human recombit protein fragment corresponding to amino acids 100-362 of human LHX1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.