Learn More
LHX2 Mouse anti-Human, Mouse, Clone: OTI2E8, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF810304
Catalog No. 50-166-9997
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator. The protein can recapitulate or rescue phenotypes in Drosophila caused by a related protein, suggesting conservation of function during evolution.
Specifications
Specifications
LHX2
Monoclonal
Unconjugated
LHX2
Lyophilized
LHX2
Human recombit protein fragment corresponding to amino acids 220-406 of human LHX2 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Mouse
Western Blot
OTI2E8
PBS with 8% trehalose and no preservative; pH 7.3
P50458, Q9Z0S2
LH2, hLhx2
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
16870, 9355