Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LHX5, Rabbit anti-Human,Mouse,Rat, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP280517
Description
LHX5 Polyclonal antibody specifically detects LHX5 in Human, Mouse, Rat Samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
LHX5 | |
Unconjugated | |
Lyophilized from whole serum | |
LIM homeobox 5, LIM homeobox protein 5, LIM/homeobox protein Lhx5, MGC129689 | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
64211 |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Western Blot 1:300 - 1:2000, Immunohistochemistry 1:300 - 1:2000, Immunohistochemistry-Paraffin 1:300 - 1:2000 | |
Unpurified | |
LHX5 | |
A synthetic peptide from aa region 150-200 of human LHX5 conjugated to blue carrier protein was used as the antigen. The peptide is homologous with rat and mouse sequences. | |
0.1 mL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Polyclonal | |
Human, Mouse, Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title