Learn More
LHX5 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 215671AP
Catalog No. 50-173-3488
Add to cart
Description
Description
This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator and be involved in the control of differentiation and development of the forebrain. In mice, this protein is essential for the regulation of precursor cell proliferation and the control of neuronal differentiation and migration during hippocampal development. This protein is involved in learning and motor functions in adult mice.
Specifications
Specifications
LHX5
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H2C1
LHX5, LIM homeobox 5, LIM homeobox protein 5, LIM/homeobox protein Lhx5
Rabbit
IgG
150 μL
-20°C
Primary
64211
Western Blot
Unconjugated
LHX5
Liquid
LHX5
LHX5 Fusion Protein Ag16081
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human