Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIAT1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP321482
Description
LIAT1 Polyclonal antibody specifically detects LIAT1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
LIAT1 | |
Polyclonal | |
Western Blot 1:500-1:2000, Immunohistochemistry, Immunoprecipitation 1-5uL/mg lysate, Immunohistochemistry-Paraffin 1:50-1:200 | |
chromosome 17 open reading frame 97, hypothetical protein LOC400566 | |
Produced in rabbits immunized with E. coli-derived Human LIAT1 fragment | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH7.0 | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
400566 | |
Store at 4°C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction