Learn More
MilliporeSigma™ LiChroCART™ HPLC Cartridge, 25-4
Available in a variety of stationary phases that function in normal and/or reverse phase. MilliporeSigma LiChroCART™ HPLC Cartridge, 25-4 comes with customized packing (please specify packing material) and is 25mmL x 4mmI.D.
$200.18 - $305.00
Specifications
Column Type LiChroCART™ HPLC cartridge
Length (Metric) 25mm
Diameter (Metric) Inner 4mm
Sorbent Customized (specify packing material)
View More Specs
Products 2
Catalog Number Mfr. No. Stationary Phase Price Quantity & Availability  
Catalog Number Mfr. No. Stationary Phase Price Quantity & Availability  
M1501727075 View Documents MilliporeSigma™
1.50172.7075
DIOL5 Each for $200.18
Add to cart
M1501727185 View Documents MilliporeSigma™
1.50172.7185
Each for $305.00
Add to cart
Specifications
Specifications
LiChroCART™ HPLC cartridge
25mm
4mm
Customized (specify packing material)