Learn More
MilliporeSigma™ LiChroCART™ HPLC Cartridge, 250-2
Available in a variety of stationary phases that function in normal and/or reverse phase. MilliporeSigma LiChroCART™ HPLC Cartridge, 250-2 comes with customized packing (please specify packing material) and is 250mmL x 2mmI.D.
$718.71 - $1060.02
Specifications
Column Type LiChroCART™ HPLC cartridge
Length (Metric) 250mm
Diameter (Metric) Inner 2mm
Sorbent Customized (specify packing material)
View More Specs
Products 6
Catalog Number Mfr. No. Stationary Phase Mobile Phase Compatibility Price Quantity & Availability  
Catalog Number Mfr. No. Stationary Phase Mobile Phase Compatibility Price Quantity & Availability  
M1501907075 View Documents MilliporeSigma™
1.50190.7075
DIOL5 RP (reversed-phase, when in mode) & NP (normal-phase) Each for $731.69
Add to cart
M1501907087 View Documents MilliporeSigma™
1.50190.7087
Each for $735.00
Add to cart
M1501907109 View Documents MilliporeSigma™
1.50190.7109
60SI NP (normal-phase) Each for $718.71
Add to cart
M1501907175 View Documents MilliporeSigma™
1.50190.7175
SI5 NP (normal-phase) Each for $828.82
Add to cart
M1501907177 View Documents MilliporeSigma™
1.50190.7177
NH2 RP (reversed-phase, when in mode) & NP (normal-phase) Each for $874.84
Add to cart
M1501907184 View Documents MilliporeSigma™
1.50190.7184
RP18 RP (reversed-phase) Each for $1,060.02
Add to cart
Specifications
Specifications
LiChroCART™ HPLC cartridge
250mm
2mm
Customized (specify packing material)