Learn More
MilliporeSigma™ LiChroCART™ HPLC Cartridge, 250-4.6
Available in a variety of stationary phases that function in normal and/or reverse phase. MilliporeSigma™ LiChroCART™ HPLC Cartridge, 250-4.6 comes with customized packing (please specify packing material) and is 250mmL x 4.6mmI.D.
Supplier:  MilliporeSigma™ 1.51431.7084
Catalog No. M1514317084
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Specifications
Specifications
LiChroCART™ HPLC Cartridge
4.6 mm
RP (reversed-phase)
Customized (specify packing material)
250 mm
Product Suggestions
Product Suggestions