Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LiChropur™ N-Methyl-N-(trimethylsilyl)trifluoroacetamide with 1 trimethylchlorosilane, ≥97%, MilliporeSigma™ Supelco™
Silylation can be performed by adding N-Methyl-N-(trimethylsilyl)trifluoroacetamide (MTFA) during analysis of fluorometabolites via gas chromatography-mass spectrometry (GC-MS).
Supplier: MilliporeSigma Supelco 6947810X1MLF
Specifications
24589-78-4 | |
Organic Standard | |
C6H12F3 NOSi | |
199.25 | |
GC Derivatization |
10 x 1 mL | |
LiChropur | |
MFCD00000411 | |
≥97% | |
26°C - closed cup |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany
Spot an opportunity for improvement?
Provide Content Correction