Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LiChropur™ N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 98.5%, MilliporeSigma™ Supelco™
N-Methyl-N-(trimethylsilyl)trifluoroacetamide (MSTFA) is a silylating reagent.
$152.00 - $456.00
Specifications
CAS | 24589-78-4 |
---|---|
Type | Organic Standard |
Product Line | LiChropur |
Linear Formula | CF3 CON(CH3)Si(CH3)3 |
MDL Number | MFCD00000411 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
11-101-9198
|
MilliporeSigma Supelco
694795ML |
5 mL |
Each for $152.00
|
|
|||||
11-101-8706
|
MilliporeSigma Supelco
6947910X1ML |
10 x 1 mL |
Each for $238.00
|
|
|||||
11-101-8278
|
MilliporeSigma Supelco
6947925ML |
25 mL |
Each for $456.00
|
|
|||||
Specifications
24589-78-4 | |
LiChropur | |
MFCD00000411 | |
199.25 | |
GC Derivatization | |
>1 (vs. air) | |
130°C to 132°C |
Organic Standard | |
CF3 CON(CH3)Si(CH3)3 | |
N-Trimethylsilyl-N-methyl trifluoroacetamide; MSTFA | |
≥98.5% | |
8.8 mmHg (27°C) | |
1.075 g/mL (at 25°C) | |
26°C - closed cup |
Safety and Handling
Recommended Storage : 2°C to 8°C
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title