Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ N-Methyl-N-trimethylsilylheptafluorobutyramide, 90%, MilliporeSigma™ Supelco™

$94.90 - $260.50

Specifications

CAS 53296-64-3
Product Line LiChropur
Percent Purity ≥90% (GC)
Density 1.254 g/mL (at 25°C)
Grade For GC derivatization
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-9233
View Documents
MilliporeSigma Supelco
694841ML
1 mL
Each for $94.90
Add to cart
 
11-101-8336
View Documents
MilliporeSigma Supelco
694845ML
5 mL
Each for $260.50
Add to cart
 
Description

Description

Specifications

Specifications

53296-64-3
≥90% (GC)
For GC derivatization
LiChropur
1.254 g/mL (at 25°C)
CF3 CF2 CF2 CON(CH3)Si(CH3)3
SDS
Product Certifications

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit