Learn More
MilliporeSigma™ LiChrospher™ 60 RP-select B LiChroCART™ HPLC Cartridge, 250-4
Well suited for the separation of neutral, acidic and weak basic compounds. MilliporeSigma LiChrospher™ 60 RP-select B LiChroCART™ HPLC Cartridge, 250-4 features a reliable and versatile traditionally produced spherical silica gel carrier that
$692.79 - $918.14
Specifications
Column Type LiChroCART™ HPLC cartridge
Sorbent Silica gel
Length (Metric) 250mm
Diameter (Metric) Inner 4mm
View More Specs
Products 2
Catalog Number Mfr. No. Particle Size Price Quantity & Availability  
Catalog Number Mfr. No. Particle Size Price Quantity & Availability  
M1507420001 View Documents MilliporeSigma™
1.50742.0001
10μm Each for $692.79
Add to cart
M1508390001 View Documents MilliporeSigma™
1.50839.0001
5μm Each for $918.14
Add to cart
Specifications
Specifications
LiChroCART™ HPLC cartridge
Silica gel
250mm
4mm