Learn More
MilliporeSigma™ Supelco™ LiChrospher™ CN HPLC Column, 5 µm Particle Size
Use guard cartridge holder 54987
Manufacturer:  MilliporeSigma™ Supelco™ 54788
Catalog No. 11-100-3267
Add to cart
Specifications
Specifications
No
Reversed Phase, Normal Phase, HILIC
25 cm
100 Å
HPLC
LiChrospher
Cyano Phase/Cyanopropyl
4.6 mm
5 μm
350 m2/g
Cyano
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
LiChrospher is a registered trademark of Merck KGaA, Darmstadt, Germany