Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Lid for Polystyrene Plant Tissue Container

For aseptic germination of seeds or spores.

$570.00 - $2060.00

Specifications

Sterility Non-sterile
Material Polystyrene
Color Clear
Description Closure (accessory)
View More Specs

Products 2
Catalog Number Mfr. No. For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) Price Quantity & Availability  
07-000-963
View Documents Promotion Details
Greiner Bio-One
960178
Plant tissue culture containers (175mL)
Case of 315 for $2,060.00
Add to cart
 
07-000-967
View Documents Promotion Details
Greiner Bio-One
967178
Plant tissue culture containers (190mL)
Case of 50 for $570.00
Add to cart
 
Description

Description

  • Closure caps
  • For use with polystyrene plant tissue culture container (Part No: 960177)
Specifications

Specifications

Non-sterile
Polystyrene
Clear
Closure (accessory)
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit