Learn More
LIF Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Manufacturer:  R&D Systems AF449SP
Catalog No. AF449SP
Add to cart
This item is not returnable. View return policy
Description
Description
LIF Polyclonal antibody AF449SP specifically detects LIF in Mouse samples. It is validated for Neutralization, Western Blotting
Specifications
Specifications
LIF
LYOPH
Western Blot 0.1 ug/mL, Neutralization 10-60 ng/mL
Affinity Purified
LIF
E. coli-derived recombinant mouse LIF
Antigen Affinity-purified
RUO
Primary
3976
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot, Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDA, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergi
Goat
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse LIF in direct ELISAs and Western blots. In direct ELISAs and Western blots (non-reducing conditions), less than 5% cross-reactivity with recombinant human (rh) LIF is observed. In direct ELISAs, no cross-reactivity with rhCNTF, recombinant mouse (rm) G-CSF, rmIL-6, and rmOSM is observed.
Mouse