Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nasco Life/form™ Basic Sanitary CPR Dog



Click to view available options
CPR Dog
Description
- The Life/form™ Basic Sanitary CPR Dog provides each student with their own sanitary muzzle and nosepiece with a one-way valve.
- The muzzles may be disinfected and reused, while the nosepieces and lower airways are disposable.
- In addition to practice in assisted breathing, students may practice the appropriate rate, pressure, and position of cardiac massage and coordinate the respiratory cardiovascular functions.
- A visible chest rise appears when correct ventilations are applied.
- Includes soft carrying case, six muzzles, 10 nosepieces, and three lower disposable airways.
Specifications
Specifications
Product Type | Basic Sanitary CPR Dog |
For Use With (Application) | Animal Medical Practice |
Color | Black |
Includes | Soft carrying case, six muzzles, 10 nosepieces, and three lower disposable airways. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction