Thermo Scientific™ Life Science Analyzer: BioMate™ 3S Spectrophotometer

Your busy life science laboratory demands performance and flexibility — count on the Thermo Scientific™ BioMate 3S UV-Visible spectrophotometer to deliver.

Manufacturer: Thermo Scientific™ 840208400

 View more versions of this product

Catalog No. 14-386-504

  Check Availability
Add to Cart

    Specifications

    Drift <0.0005A/hr
    Data Interval for Scanning 0.1, 0.2, 0.5, 1.0, 2.0, 5.0 nm
    Description BioMate 3S Spectrophotometer with printer
    Detector Type Dual Silicon Photodiodes
    Photometric Display -0.1A - 4.0A; -1.5 - 125%T; ±9999C
    Photometric Linearity Up to 3.5A at 260 nm
    Scan Speed Up to 3600 nm/min
    Connections USB Type A port for USB memory device (front panel)
    USB Type B port for optional computer connectivity (rear panel)
    USB Type A port for external printer (rear panel)
    Container Type Single cell holder and automated 6-cell changer standard
    Display Graphical with LCD backlight; 9.7 x 7.1 cm (3.8 x 2.8 in)
    Electrical Requirements Selected automatically, 100-240V; 50-60Hz
    Includes USB cable, USB memory device
    Keypad Sealed membrane with tactile response keys
    Photometric Accuracy Instrument  ±0.005A at 1.0A
    Printer Internal printer:
    40 column internal (text and graphics)
    Included with PNs 840-208100 and 840-209800
    Optional accessory for PNs 840-209700

    External Printer Support:
    Expernal printer with USB connection and HP PCL 3.0 or higher printer language support
    Wavelength Repeatability  ±0.5 nm
    Type BioMate 3S Spectrophotometer with printer
    In Germany, France, and Spain use Catalog #840-210000
    Weight (English) 19 lb.
    Weight (Metric) 8.6 kg
    Width (English) 13 in.
    Width (Metric) 33 cm
    Depth (English) 16 in.
    Depth (Metric) 41 cm
    Height (English) 9 in.
    Height (Metric) 23 cm
    Interface RS-232-C Bidirectional
    Lamp Xenon Flash Lamp (5 years, 3 years guaranteed)
    Noise <0.00025 at 0.0A;
    <0.00050 at 1.0A;
    <0.00080 at 2.0A;
    MS at 260 nm
    Optical Design Dual Beam with Internal Reference Detector
    Spectral Bandwidth 1.8 nm
    Stray Light <0.08%T at 220, 340 nm (Nal, NaNO2); <1.0%T 198-200 nm (KCI)
    Wavelength Accuracy ±1.0 nm
    Wavelength Range 190 - 1,100 nm

    Your busy life science laboratory demands performance and flexibility — count on the Thermo Scientific™ BioMate 3S UV-Visible spectrophotometer to deliver. With stray light and noise specifications comparable to instruments two or three times the price, the new, compact  BioMate 3S spectrophotometer saves you valuable bench space. The BioMate 3S spectrophotometer offers pre-programmed assay methods for nucleic acid and protein concentration as well as cell culture measurements.

    The BioMate 3S spectrophotometer is a highly configurable instrument, adaptable to the needs of the lab. A variety of accessories adds convenience and provides Peltier temperature control, automation, and fiber optic sampling.

    Hardware Designed for Performance

    • Improved, 1.8nm spectral bandwidth is compliant with worldwide Pharmacopeia requirements, allowing research quality measurements with routine simplicity
    • Patented optical design provides a compact, high-performance dual-beam system with very few moving parts
    • Xenon lamp gives balanced light over the full instrument wavelength range of 190 to 1100nm
    • Dual-beam optical system includes an internal reference detector and offers significant advantages over a single beam or diode array instruments, including compensation for lamp intensity changes, better performance for kinetics, and better long-term stability

    Software Enhanced Flexibility

    • Embedded software provides pre-programmed assays for RNA/DNA concentration or purity estimation, protein concentrations, cell growth, kinetics, or routine measurements
    • For more intense applications, optional application software programs allow more sophisticated data collection, analysis, and reporting

    Time-Saving Bioanalysis Software

    • Pre-programmed assays deliver fast, accurate results
    • Built-in assays for nucleic acid concentration and purity estimations, protein concentration, and cell growth at 600nm
    • Flexible Oligo calculator feature for calculating molecular weight, theoretical Tm and Oligo concentration factor
    • Other standard applications include single and multicell kinetics, wavelength scanning, and user-defined fixed wavelength measurements

    BioMate 3S Spectrophotometer

    • Unit is ideal for DNA/RNA and protein analysis
    • Optional built-in 3in. (7.6cm) graphical printer shows test parameters, graph, and data
    • Optional automatic six-position cell holder speeds up cell growth measurements or standard curve preparations

    Pre-programmed life science assays include:

    • Nucleic acid ratio and concentration (260/280 and 260/230)
    • Direct protein concentration at 280nm and 205nm
    • Coomassie™/Bradford (Standard and Micro)
    • Lowry
    • BCA
    • Pierce Micro-BCA
    • Pierce 660nm Protein Assay
    • Cell growth (with scaling factor)
    • Kinetics

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit