Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Discovery Education™ Life Science: The Basics

Introduce young students to a wide variety of life science topics.
Supplier: Discovery Education™ DC879163
Description
Video segments are drawn from an award-winning series.
- Chaptered into navigable units for quick access to desired content segments
- Six segments highlight techniques for teaching with inquiry, activators, investigation and summarizers
- Highlights correlation to national education standards
- Includes teacher's guide: The Use of Multimedia in the Classroom
Grades K to 2
- Ecology: Introduction
- Temperate Rain Forest
- Polar Ice
- Plants
- The Lives of Butterflies
- Mammals: Introduction
- Reptiles: Introduction
- Frogs & Toads
- Invertebrates: Introduction
- Molds
- Human Biology: Introduction
- Nutrition
- The Cell
- Bacteria: Introduction
- The Rain Forest and the Desert
- Coral Ecosystems
- Organisms: Undersea Discoveries
- Birds and Insects
- Symbiosis
- How Plants Adapt
- How Bones Develop
- Skin
- Food & Digestion
- The Genetics of Twins
Specifications
3 to 5 | |
VHS | |
Marc Record, Teacher's Guide |
2005 | |
180 min. | |
Life Science: the Basics Grades 3-5 VHS |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title