Learn More
Lifescan SureStep™ Pro System Controls
Manufacturer:  LifeScan 53885079525
 View more versions of this product
Catalog No. 23-029295
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Low glucose control solution
40mg/dL
Any SureStep™ brand Blood Glucose meter with SureStep™Pro™ test strips
25 Pack
Low
Test strips to check that the meter and test strips are, working together properly
30°C
Safety and Handling
ShelfLife : 3 Months