Lifescan SureStep™ Pro System Controls

Manufacturer: LifeScan 53885085002

 View more versions of this product

Catalog No. 23-029-350

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type High/Low glucose control solution
    Shelf Life 3 Months
    Storage Requirements 30°C
    Quantity 2 Pack,2 Pack
    Level High, Low
    Concentration 345/40mg/dL
    For Use With (Application) Test strips to check that the meter and test strips are, working together properly
    For Use With (Equipment) Any SureStep™ brand Blood Glucose meter with SureStep™Pro™ test strips

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit