Lifescan SureStep™ Pro System Test Strips
SureStep Pro Test Strips are for use with SureStep Brand Blood Glucose Meters for quantitatively measuring glucose levels in whole blood.
SureStep Pro Test Strips may be used to test capillary, venous, arterial, and neonatal blood samples.
Required Accessories:
  • SureStep Brand Blood Glucose Meter and applicable operator's guide.
  • Single–use lancing device, such as Lifescan Unisik' 2 or equivalent (If obtaining capillary sample)
  • Products 1
  • Description & Specifications
Products
Catalog Number Mfr. No. Description Price Quantity Availability  
23-029296
Lifescan
53885079750
Test Strips for SureStep meter
N/A N/A N/A  
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
Descriptions & Specifications
Related Products