Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ LIFR alpha Antibody Pair [HRP]

Polyclonal/Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2796485PLATES

 View more versions of this product

Catalog No. NB010336


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes an antibody specific for Rat LIFR alpha.
Specifications

Specifications

39.06 pg/mL
Conjugated
3977
Antibody Pairs
39.06-2500 pg/mL
Storage is content dependent.
Rabbit/Mouse
Rat
LIFR
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2500 pg/mL is recommended.
HRP
Sandwich ELISA
  • Standard: Each vial contains 270 ng of recombinant Rat LIFR alpha.
  • Capture Antibody: 0.5 mg/mL of mouse anti-Rat LIFR alpha monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-LIFR alpha polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • RUO
    This solid phase sandwich ELISA utilizes an antibody specific for Rat LIFR alpha.
    LIFR alpha
    5 Plates
    Sandwich ELISA
    CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF receptor, LIF-R, SJS2, STWS, SWS
    SDS
    Documents
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit