Learn More
ACROBiosystems Recombinant Protein;Human LIF R / CD118 Protein, Fc Tag;HEK293;200UG;LIR,LIFR,CD118,FLJ98106,FLJ99923,LIF-R,SJS2,STWS,SWS
Supplier:  ACROBiosystems LIRH4252200UG
Encompass_Preferred
Recombinant Protein;Human LIF R / CD118 Protein, Fc Tag;200UG;Human LIF R, Fc Tag (LIR-H4252) is expressed from human 293 cells (HEK293). It contains AA Gln 45 - Ser 833 (Accession # NP_001121143).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 115.5 kDa. The protein migrates as 135-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LIF, Tag Free (Cat. No. LIF-H521b) at 0.2μg/mL (100 μL/well) can bind Human LIF R, Fc Tag (Cat. No. LIR-H4252) with a linear range of 7.8-31.25 ng/mL (QC tested).
Catalog No. 50-165-7700