Thermo Scientific™ LifterSlip™ mSeries Cover Slips for Microarray Slides Available on GSA/VA Contract for Federal Government customers only.
Reduce the variation in hybridization fluid volume with the superior surface quality and flatness of Thermo Scientific™ LifterSlip™ mSeries Cover Slips for Microarray Slides.
Manufacturer: Thermo Scientific™ 22X25IM5226001LS
 View more versions of this product
Catalog No. 22-037-220
  Check Availability
Add to Cart
    Specifications
    Thickness (Metric) 1mm
    Size (Metric) 25 x 22mm
    Volume (Metric) 20µL
    Description mSeries LifterSlip Cover Slips for Microarray Slides
    Reduce the variation in hybridization fluid volume across the microarray surface with the superior surface quality and flatness of Thermo Scientific™ LifterSlip™ mSeries Cover Slips for Microarray Slides. These slides are made of 1mm thick microarray quality, ultra flat glass.
    • Feature precise ink-printed raised edges to provide consistent dispersal and easy injection of hybridization solution
    • Manufactured without adhesives to prevent leaching at high temperatures or extended hybridization times
  • Description & Specifications
Related Products
Product Content Feedback