Learn More
Thermo Scientific™ LifterSlip™ mSeries Cover Slips for Microarray Slides
Reduce the variation in hybridization fluid volume with the superior surface quality and flatness of Thermo Scientific™ LifterSlip™ mSeries Cover Slips for Microarray Slides.
Specifications
Description mSeries LifterSlip Cover Slips for Microarray Slides
View More Specs
Products 1
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
22-037-223
View Documents
Thermo Scientific™
25X60IM5439001LS
55µL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Reduce the variation in hybridization fluid volume across the microarray surface with the superior surface quality and flatness of Thermo Scientific™ LifterSlip™ mSeries Cover Slips for Microarray Slides. These slides are made of 1mm thick microarray quality, ultra flat glass.
  • Precise ink-printed raised edges to provide consistent dispersal and easy injection of hybridization solution
  • Without adhesives to prevent leaching at high temperatures or extended hybridization times
Specifications
Specifications
mSeries LifterSlip Cover Slips for Microarray Slides