Learn More
Thermo Scientific™ LifterSlips™ Cover Slips for Microarray Slides Available on GSA/VA Contract for Federal Government customers only.
Accommodate various hybridization solution volumes with Thermo Scientific™ LifterSlip™ Cover Slips for Microarray Slides.
Manufacturer:  Thermo Scientific™ 22X50I24711001LS
 View more versions of this product
Catalog No. 22-035-804
  Check Availability
Add to cart
Specifications
Description
Accommodate various hybridization solution volumes with Thermo Scientific™ LifterSlip™ Cover Slips for Microarray Slides. This product prevents glass-to-glass contact between cover slip and microarray.
  • Standard thickness number 2 (tolerance 0.17–0.25mm)
  • Printed raised edges enable easy injection and uniform dispersal of hybridization solution under prepositioned cover slips
  • Capillary attraction wicks solution under cover slip and keeps the LifterSlip in place
Description & Specifications
Specifications
50 x 22mm
50mm
0.17 to 0.25mm
47 oz.
22mm
60µL