Learn More
Thermo Scientific™ LifterSlips™ Cover Slips for Microarray Slides
Accommodate various hybridization solution volumes with Thermo Scientific™ LifterSlip™ Cover Slips for Microarray Slides.
Products 6
Catalog Number Mfr. No. Length (Metric) Width (Metric) Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Width (Metric) Volume (Metric) Price Quantity & Availability  
22-035-804
View Documents
Thermo Scientific™
22X50I24711001LS
50mm 22mm 60µL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
22-035-809
View Documents
Thermo Scientific™
25X60I247891LS
60mm 25mm 75µL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
22-035-803
View Documents
Thermo Scientific™
22X40I24710001LS
40mm 22mm 50µL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
22-035-800
View Documents
Thermo Scientific™
22X22I247881LS
22mm 22mm 26µL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
22-035-802
View Documents
Thermo Scientific™
22X30I24734001LS
30mm 22mm 42µL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
22-035-805
View Documents
Thermo Scientific™
22X60I24861001LS
60mm 22mm 75µL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Accommodate various hybridization solution volumes with Thermo Scientific™ LifterSlip™ Cover Slips for Microarray Slides. This product prevents glass-to-glass contact between cover slip and microarray.
  • Standard thickness number 2 (tolerance 0.17–0.25mm)
  • Printed raised edges enable easy injection and uniform dispersal of hybridization solution under prepositioned cover slips
  • Capillary attraction wicks solution under cover slip and keeps the LifterSlip in place