Learn More
Cen-Med Light Blue Disposable Short Sleeve V-Neck Scrub Top with 3 Pockets
Single use disposable scrub top provides superior protection against contaminants and prevents bodily fluids from making contact with skin
$268.73
Specifications
Color Light Blue
Material SMS Non-Woven
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
21-520-211
View Documents
Cen-Med
C3007011100
Small
Case of 100 for $268.73
Add to cart
21-520-212
View Documents
Cen-Med
C3007012100
Medium
Case of 100 for $268.73
Add to cart
21-520-213
View Documents
Cen-Med
C3007013100
Large
Case of 100 for $268.73
Add to cart
21-520-214
View Documents
Cen-Med
C3007014100
X-Large
Case of 100 for $268.73
Add to cart
Description
Description
  • Optimal coverage, comfort and breathability
  • Ease of range of movement
  • Hypoallergenic
  • SMS Non Woven Material
  • Anti-Static
  • Latex Free
  • 45 g
Specifications
Specifications
Light Blue
SMS Non-Woven